missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Adipate 99.0+%, TCI America™

Catalog No. A0706500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0706500ML 500 mL
A070625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0706500ML Supplier TCI America Supplier No. A0706500ML

Chemical Identifiers

CAS 141-04-8
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.358
MDL Number MFCD00053722
InChI Key RDOFJDLLWVCMRU-UHFFFAOYSA-N
Synonym diisobutyl adipate, isobutyl adipate, diba, ftaflex diba, plasthall diba, bis 2-methylpropyl hexanedioate, hexanedioic acid, bis 2-methylpropyl ester, adipic acid, diisobutyl ester, diisobutyl hexanedioate, adipic acid bis 2-methylpropyl ester
PubChem CID 8831
ChEBI CHEBI:34708
IUPAC Name bis(2-methylpropyl) hexanedioate
SMILES CC(C)COC(=O)CCCCC(=O)OCC(C)C

Specifications

Color Colorless
Boiling Point 293°C
Quantity 500 mL
Formula Weight 258.36
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisobutyl Adipate
EINECSNumber (2)-0861&(2)-0879
RTECSNumber AV1480000
TSCA Yes