missing translation for 'onlineSavingsMsg'
Learn More

Diiodomethane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. D061025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D061025G 25 g
D0610100G 100 g
D0610500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D061025G Supplier TCI America Supplier No. D061025G

Chemical Identifiers

CAS 75-11-6
Molecular Formula CH2I2
Molecular Weight (g/mol) 267.84
MDL Number MFCD00001079
InChI Key NZZFYRREKKOMAT-UHFFFAOYSA-N
Synonym methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
PubChem CID 6346
IUPAC Name diiodomethane
SMILES ICI

Specifications

Color Yellow
Boiling Point 181°C
Quantity 25 g
UN Number 1760
Formula Weight 267.84
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diiodomethane (stabilized with Copper chip)
EINECSNumber (2)-0043
RTECSNumber PA8575000
TSCA Yes