Learn More
Diiodomethane, 99+%, stabilized with silver wire
CAS: 75-11-6 | CH2I2 | 267.84 g/mol
Supplier: Thermo Scientific Chemicals 169830025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Diiodomethane | |
| 6.0°C | |
| 3.3200g/mL | |
| >110°C | |
| 99% min. (GC) | |
| CH2I2 | |
| CH2I2 | |
| 2.5 kg | |
| 09,170; 11,182; 17,116 | |
| 15,6137 | |
| Solubility in water: insoluble. Other solubilities: miscible with alcohol,propanol,isopropanol,,hexane,cyclohexane,ether,chloroform and benzene | |
| ICI | |
| 267.84 | |
| 6346 | |
| 267.83 | |
| Liquid |
| 75-11-6,7440-22-4 | |
| Red to Yellow | |
| 181.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.737 | |
| MFCD00001079 | |
| 01,71 | |
| 3.32 | |
| methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane | |
| NZZFYRREKKOMAT-UHFFFAOYSA-N | |
| diiodomethane | |
| Silver wire | |
| 2.80 mPa.s (20°C) | |
| 99+% |
Chemical Identifiers
| 75-11-6 | |
| 267.84 | |
| NZZFYRREKKOMAT-UHFFFAOYSA-N | |
| 6346 | |
| ICI |
| CH2I2 | |
| MFCD00001079 | |
| methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane | |
| diiodomethane |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 200-841-5
RUO – Research Use Only