Learn More
Diiodomethane, 99%, stab.
CAS: 75-11-6 | CH2I2 | 267.84 g/mol
Supplier: Thermo Scientific Chemicals A1545722
Description
Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.
Solubility
Miscible with alcohol and ether. Slightly miscible with water.
Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with alkali metal salts, strong oxidizing agents, strong bases, metals, lithium, potassium, sodium, sodium oxides and alloys.
Specifications
| Diiodomethane | |
| 75-11-6 | |
| 3.322 | |
| None | |
| CH2I2 | |
| MFCD00001079 | |
| UN2810 | |
| Light sensitive | |
| methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane | |
| NZZFYRREKKOMAT-UHFFFAOYSA-N | |
| diiodomethane | |
| 6346 | |
| 99% |
| stabilized | |
| 5°C to 6°C | |
| 180°C to 181°C (decomposition) | |
| 99%, stab. | |
| 1.74 | |
| 100 g | |
| 1696892 | |
| 14,6066 | |
| Miscible with alcohol and ether. Slightly miscible with water. | |
| ICI | |
| 267.84 | |
| 267.84 | |
| Liquid |
Chemical Identifiers
| 75-11-6 | |
| 267.84 | |
| NZZFYRREKKOMAT-UHFFFAOYSA-N | |
| 6346 | |
| ICI |
| CH2I2 | |
| MFCD00001079 | |
| methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane | |
| diiodomethane |
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H301-H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 200-841-5
RTECSNumber : PA8575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only