missing translation for 'onlineSavingsMsg'
Learn More

Diiodomethane, 99%, stab.

Catalog No. AAA1545714
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
500 g
1000 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1545714 25 g
AAA1545718 50 g
AAA1545722 100 g
AAA1545730 250 g
AAA1545736 500 g
AAA154570B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. AAA1545714 Supplier Thermo Scientific Chemicals Supplier No. A1545714

CAS: 75-11-6 | CH2I2 | 267.84 g/mol

Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.

Solubility
Miscible with alcohol and ether. Slightly miscible with water.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with alkali metal salts, strong oxidizing agents, strong bases, metals, lithium, potassium, sodium, sodium oxides and alloys.

Chemical Identifiers

CAS 75-11-6
Molecular Formula CH2I2
Molecular Weight (g/mol) 267.84
MDL Number MFCD00001079
InChI Key NZZFYRREKKOMAT-UHFFFAOYSA-N
Synonym methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
PubChem CID 6346
IUPAC Name diiodomethane
SMILES ICI

Specifications

Melting Point 5°C to 6°C
Density 3.322
Boiling Point 180°C to 181°C (decomposition)
Flash Point None
Assay Percent Range 99%, stab.
Refractive Index 1.74
Quantity 25 g
UN Number UN2810
Beilstein 1696892
Sensitivity Light sensitive
Merck Index 14,6066
Solubility Information Miscible with alcohol and ether. Slightly miscible with water.
Formula Weight 267.84
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diiodomethane, stabilized
Show More Show Less
Hazard Category H301-H315-H318-H335
Hazard Statement GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 200-841-5
RTECSNumber PA8575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only