missing translation for 'onlineSavingsMsg'
Learn More

Diiodo(p-cymene)ruthenium(II) dimer

Catalog No. p-7052236
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4688603 1 g
AA4688606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4688603 Supplier Thermo Scientific Chemicals Supplier No. 04688603
Only null left

CAS: 90614-07-6

Diiodo(p-cymene)ruthenium(II) dimer can be applied as a catalyst for many reactions such as ruthenium-catalyzed hydrogenation, preparation of Walphos-type ferrocenyl-aryl diphosphine ligands, ruthenium-catalyzed synthesis reactions, and enantioselective hydrogenation of ketones and olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90614-07-6
MDL Number MFCD08064027
Synonym Di-μ-iodobis[(p-cymene)iodoruthenium(II)]

Specifications

Color Brown-Yellow
Quantity 1 g
Sensitivity Light and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 978.2
Physical Form Crystalline Solid
Chemical Name or Material Diiodo(p-cymene)ruthenium(II) dimer
Hazard Category H302-H319-H341-H500
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only