missing translation for 'onlineSavingsMsg'
Learn More

Dihydroxybis(ammonium lactato)titanium(IV), 50% w/w aq. soln.

Water soluble titanium complex | CAS: 65104-06-5

Supplier:  Thermo Scientific Chemicals 042676A1

 View more versions of this product

Catalog No. AA42676A1

Only null left
Explore available promotions

Description

Description

Dihydroxybis(ammonium lactato)titanium(IV) is used in the production of water soluble titanium complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dihydroxybis (ammonium lactato)titanium (IV)
Yellow
47°C (118°F)
MFCD00072736
UN1993
Miscible with water.
50% w/w aq. soln.
65104-06-5
1.21 g/mL
1.438
1 kg
Titanium(IV) bis(ammonium lactato)dihydroxide
294.1
Liquid

Chemical Identifiers

65104-06-5
MFCD00072736
Titanium(IV) bis(ammonium lactato)dihydroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c

H226-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 247-428-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only