missing translation for 'onlineSavingsMsg'
Learn More

Dihydroxybis(ammonium lactato)titanium(IV), 50% w/w aq. soln.

Catalog No. AA42676A7
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42676A7 5 kg
AA4267630 250 g
AA42676A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42676A7 Supplier Thermo Scientific Chemicals Supplier No. 042676A7
Only null left

Water soluble titanium complex | CAS: 65104-06-5

Dihydroxybis(ammonium lactato)titanium(IV) is used in the production of water soluble titanium complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65104-06-5
MDL Number MFCD00072736
Synonym Titanium(IV) bis(ammonium lactato)dihydroxide

Specifications

Color Yellow
Density 1.21 g/mL
Flash Point 47°C (118°F)
Refractive Index 1.438
Quantity 5 kg
UN Number UN1993
Solubility Information Miscible with water.
Formula Weight 294.1
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Dihydroxybis (ammonium lactato)titanium (IV)
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 247-428-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only