missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydroxybis(ammonium lactato)titanium(IV), 50% w/w aq. soln.
Description
Dihydroxybis(ammonium lactato)titanium(IV) is used in the production of water soluble titanium complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Density | 1.21 g/mL |
| Flash Point | 47°C (118°F) |
| Refractive Index | 1.438 |
| Quantity | 250 g |
| UN Number | UN1993 |
| Solubility Information | Miscible with water. |
| Formula Weight | 294.1 |
| Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319 |
| Hazard Statement | GHS H Statement H226-H315-H319 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 247-428-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only