missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Dihydrorhodamine 123, Calbiochem™,

Catalog No. 3098255MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3098255MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3098255MG Supplier MilliporeSigma™ Supplier No. 3098255MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cell-permeable fluorogenic probe that is useful for the detection of reactive oxygen species (ROS) such as peroxide and peroxynitrite

Chemical Identifiers

CAS 109244-58-8
Molecular Formula C21H18N2O3
Molecular Weight (g/mol) 346.39
MDL Number MFCD04221428
InChI Key FNEZBBILNYNQGC-UHFFFAOYSA-N
Synonym dihydrorhodamine 123, dihydrorhodamine, methyl 2-3,6-diamino-9h-xanthen-9-yl benzoate, benzoic acid, 2-3,6-diamino-9h-xanthen-9-yl-, methyl ester, dihydrorhodamine-123
PubChem CID 105032
IUPAC Name methyl 2-(3,6-diamino-9H-xanthen-9-yl)benzoate
SMILES COC(=O)C1=CC=CC=C1C1C2=CC=C(N)C=C2OC2=CC(N)=CC=C12

Specifications

Color Pink-White
pH 14 (aq. soln.)
Refractive Index 1.37 (20°C, 589nm)
Quantity 5 mg
Vapor Pressure 263 hPa (20°C)
Formula Weight 346.4
Percent Purity ≥95%
Grade HPLC
Physical Form Solid
Chemical Name or Material Dihydrorhodamine 123
Recommended Storage -20°C (-4°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).