missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydrolinalool 97.0+%, TCI America™
$78.29 - $577.86
Chemical Identifiers
| CAS | 18479-51-1 |
|---|---|
| Molecular Formula | C10H20O |
| Molecular Weight (g/mol) | 156.269 |
| MDL Number | MFCD00059140 |
| InChI Key | JRTBBCBDKSRRCY-UHFFFAOYSA-N |
| Synonym | 3,7-Dimethyl-6-octen-3-ol |
| PubChem CID | 86749 |
| IUPAC Name | 3,7-dimethyloct-6-en-3-ol |
| SMILES | CCC(C)(CCC=C(C)C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D159225ML
|
TCI America
D159225ML |
25 mL |
Each for $78.29
|
|
|||||
|
D1592500ML
|
TCI America
D1592500ML |
500 mL |
Each for $577.86
|
|
|||||
Chemical Identifiers
| 18479-51-1 | |
| 156.269 | |
| JRTBBCBDKSRRCY-UHFFFAOYSA-N | |
| 86749 | |
| CCC(C)(CCC=C(C)C)O |
| C10H20O | |
| MFCD00059140 | |
| 3,7-Dimethyl-6-octen-3-ol | |
| 3,7-dimethyloct-6-en-3-ol |
Specifications
| 18479-51-1 | |
| Yellow | |
| C10H20O | |
| 25 mL | |
| JRTBBCBDKSRRCY-UHFFFAOYSA-N | |
| 3,7-dimethyloct-6-en-3-ol | |
| 86749 | |
| ≥97.0% (GC) | |
| Dihydrolinalool |
| 5°C | |
| 200°C | |
| MFCD00059140 | |
| 3,7-Dimethyl-6-octen-3-ol | |
| CCC(C)(CCC=C(C)C)O | |
| 156.269 | |
| 156.27 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0258
TSCA : Yes