missing translation for 'onlineSavingsMsg'
Learn More

Dihydrojasmone, 97%

Catalog No. AAA1929814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1929814 25 g
AAA1929822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1929814 Supplier Thermo Scientific Chemicals Supplier No. A1929814

CAS: 1128-08-1 | C11H18O | 166.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1128-08-1
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.26
MDL Number MFCD00036480
InChI Key YCIXWYOBMVNGTB-UHFFFAOYSA-N
Synonym dihydrojasmone, 3-methyl-2-pentylcyclopent-2-enone, jasmone, dihydro, 2-cyclopenten-1-one, 3-methyl-2-pentyl, 2-pentyl-3-methyl-2-cyclopenten-1-one, dihydrojasmone natural, 3-methyl-2-n-pentanyl-2-cyclopenten-1-one, 2-amyl-3-methyl-2-cyclopenten-1-one, unii-y953r7pp90, fema no. 3763
PubChem CID 62378
IUPAC Name 3-methyl-2-pentylcyclopent-2-en-1-one
SMILES CCCCCC1=C(C)CCC1=O

Specifications

Density 0.91
Boiling Point 120°C to 121°C (12 mmHg)
Flash Point 121°C (250°F)
Refractive Index 1.479
Quantity 25 g
Beilstein 1906471
Formula Weight 166.26
Percent Purity 97%
Chemical Name or Material Dihydrojasmone
EINECSNumber 214-434-5
RTECSNumber GY7302000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only