missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexahydroxyplatinate(IV), 99.9% (metals basis), Pt 61.0% min

Catalog No. p-7048626
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1105202 0.5 g
AA1105203 1 g
AA1105206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1105202 Supplier Thermo Scientific Chemicals Supplier No. 01105202
Only null left

CAS: 51850-20-5 | H8O6Pt | 299.14 g/mol

Dihydrogen hexahydroxyplatinate(IV) is used as a precursor to produce platinum dioxide (II), a catalyst of hydrogenation and hydrogenolysis of carbonyl group or C-Cl linkages in acyl chlorides. It also acts as a precursor to produce other platinum compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51850-20-5
Molecular Formula H8O6Pt
Molecular Weight (g/mol) 299.14
MDL Number MFCD00058744
InChI Key ILXDKMTYQJUXAB-UHFFFAOYSA-J
Synonym Hexahydroxyplatinic acid; Hydrogen hexahydroxyplatinate(IV)
IUPAC Name platinum(4+) dihydrogen hexahydroxide
SMILES [H+].[H+].[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Pt+4]

Specifications

Melting Point (decomposition)
Color Yellow
Quantity 0.5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 299.17
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt ≤61.0%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Dihydrogen hexahydroxyplatinate(IV)
Show More Show Less
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 257-471-2
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only