missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexafluorotitanate, 60% w/w aq. soln.

Catalog No. AA39736A1
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39736A1 1 kg
AA3973636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39736A1 Supplier Thermo Scientific Chemicals Supplier No. 039736A1

CAS: 17439-11-1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17439-11-1
MDL Number MFCD00049647
Synonym Fluorotitanic acid; Hydrogen hexafluorotitanate

Specifications

Density 1.69 g/mL
Odor Pungent
Quantity 1 kg
UN Number UN2922
Formula Weight 163.91
Concentration or Composition (by Analyte or Components) 60% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Dihydrogen hexafluorotitanate
Hazard Category H290-H301+H311+H331-H314-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 241-460-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only