missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydrogen hexafluorotitanate, 60% w/w aq. soln.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.69 g/mL |
| Odor | Pungent |
| Quantity | 500 g |
| UN Number | UN2922 |
| Formula Weight | 163.91 |
| Concentration or Composition (by Analyte or Components) | 60% w/w aq. soln. |
| Physical Form | Liquid |
| Chemical Name or Material | Dihydrogen hexafluorotitanate |
Safety and Handling
| Hazard Category | H290-H301+H311+H331-H314-H335 |
| Hazard Statement | GHS H Statement H300-H310-H330-H314-H318 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
| EINECSNumber | 241-460-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only