missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroplatinate(IV) solution, Pt 20% (cont. Pt)

Catalog No. AA40177NA
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40177NA 2 g
AA40177NJ 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40177NA Supplier Thermo Scientific Chemicals Supplier No. 040177NA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16941-12-1

Dihydrogen hexachloroplatinate(IV) is used as a catalyst in the determination of potassium and in the purification of platinum. It is also used in catalysis reactions, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and in microscopy. In the presence of copper(II) chloride, it is used to catalyze the photooxygenation of alcohols to aldehydes or ketones. It plays an important role in intramolecular hydrosilylation of an alkyne and hydrosilylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16941-12-1
MDL Number MFCD00011330

Specifications

Color Red to Brown
Physical Form Liquid
Quantity 2 g
Linear Formula H2PtCl6
UN Number UN3264
Merck Index 14,7526
Solubility Information Miscible with water,alcohol,acetone,ethyl acetate and ether.
Formula Weight 409.82 (Anhydrous)
Concentration or Composition (by Analyte or Components) Pt 20% (cont. Pt)
Chemical Name or Material Dihydrogen hexachloroplatinate(IV) solution
Hazard Category H301-H314-H317-H334-H335-H372
Hazard Statement GHS H Statement H300-H334-H372-H314-H290-H317 Fatal if swallowed. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May be corrosive to metals. May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 241-010-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only