missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroplatinate(IV) hydrate, Premion™, 99.999% (metals basis)

Catalog No. p-4781889
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4369603 1 g
AA4369606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4369603 Supplier Thermo Scientific Chemicals Supplier No. 04369603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26023-84-7 | Cl6H2Pt | 409.80 g/mol

Dihydrogen hexachloroplatinate(IV) hydrate is used in the production of platinum metal and its compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26023-84-7
Molecular Formula Cl6H2Pt
Molecular Weight (g/mol) 409.80
MDL Number MFCD00149909
InChI Key ZKOQTCCVSRSECD-UHFFFAOYSA-J
Synonym Hexachloroplatinic acid hydrate
IUPAC Name dihydrogen hexachloroplatinumtetrakis(ylium)
SMILES [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 60°C (decomposition)
Color Red to Brown
Odor Odorless
Assay Percent Range 99.999% (metals basis)
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,acetone,ethyl acetate and ether.
Formula Weight 409.82 (Anhydrous)
Physical Form Crystals
Chemical Name or Material Dihydrogen hexachloroplatinate(IV) hydrate
Hazard Category H290-H300-H314-H317-H334-H335-H372
Hazard Statement Do not breathe dust. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P234-P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 241-010-7
RTECSNumber TP1500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only