missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexachloroplatinate(IV) hexahydrate, low nitrate, Premion™, 99.95% (metals basis), Pt 37.5% min

Catalog No. p-7058586
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4281503 1 g
AA4281506 5 g
AA4281514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4281503 Supplier Thermo Scientific Chemicals Supplier No. 04281503
Only null left

CAS: 18497-13-7 | Cl6H14O6Pt | 517.89 g/mol

Dihydrogen hexachloroplatinate(IV) hexahydrate is used for the determination of potassium. It is also used for the purification of platinum by treating it with ammonium chloride flowed by heating of ammonium hexachloroplatinate with hydrogen yields elemental platinum. It also finds application in catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine color in glass and porcelain, indelible ink and microscopy. It is a precursor for the preparation of other platinum catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18497-13-7
Molecular Formula Cl6H14O6Pt
Molecular Weight (g/mol) 517.89
MDL Number MFCD00149910
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Melting Point 60°C (decomposition)
Color Red to Brown
Density 2.430 g/mL
Odor Odorless
Quantity 1 g
UN Number UN2507
Sensitivity Hygroscopic
Merck Index 14,7526
Solubility Information Soluble in water,ethanol,ether,ethyl acetate and acetone. Insoluble in nitric acid.
Formula Weight 517.91 (409.82 Anhydrous)
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 37.5% min
Physical Form Lumps
Assay (metals basis)
Chemical Name or Material Dihydrogen hexachloroplatinate (IV) hexahydrate, Low nitrate
Show More Show Less
Hazard Category H301-H314-H317-H334-H335
Hazard Statement GHS H Statement
H300-H334-H372-H314-H318-H290-H317
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CHLOROPLATINIC ACID, SOLID
EINECSNumber 241-010-7
RTECSNumber TP1500000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only