Learn More
Abcam Inc Dihydrofluorescein diacetate, Fluorescent intracellular o x idant probe, 1G
Catalog No. 502705270
Encompass
MW 418.4 Da, Purity >97%. Fluorescent intracellular oxidant probe. Redox sensor. Achieves higher intracellular concentrations resulting in brighter intracellular labelling but is less bright extracellularly than dihydrorhodamine 123 (ab145260). Oxidizes to fluorescein primarily under HO.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705270 Supplier Abcam Inc Supplier No. AB1453511G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 418.4 Da, Purity >97%. Fluorescent intracellular oxidant probe. Redox sensor. Achieves higher intracellular concentrations resulting in brighter intracellular labelling but is less bright extracellularly than dihydrorhodamine 123 (ab145260). Oxidizes to fluorescein primarily under HO.
The product is subject to the following: Abcam Restricted Use Statement