missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydroergotamine methanesulfonate
Description
Agonist at vascular serotonin receptors; partial agonist at alpha-adrenergic and dopamine D2 receptors
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Agonist at vascular serotonin receptors
- Partial agonist at alpha-adrenergic and dopamine D2 receptors
Specifications
Specifications
| Melting Point | 230°C to 235°C |
| Color | Pink to White |
| Quantity | 100 mg |
| Merck Index | 14,3174 |
| Formula Weight | 679.78 |
| Physical Form | Powder |
| Chemical Name or Material | Dihydroergotamine methanesulfonate |
Safety and Handling
| Hazard Category | H312+H332 |
| Hazard Statement | GHS H Statement H312-H332 Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c |
| EINECSNumber | 228-235-6 |
| RTECSNumber | KE7920000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only