missing translation for 'onlineSavingsMsg'
Learn More

Dihydrocoumarin, 99%

Catalog No. AAB2122922
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2122922 100 g
AAB2122936 500 g
AAB212290E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2122922 Supplier Thermo Scientific Chemicals Supplier No. B2122922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 119-84-6 | C9H8O2 | 148.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-84-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00006881
InChI Key VMUXSMXIQBNMGZ-UHFFFAOYSA-N
Synonym dihydrocoumarin, 3,4-dihydrocoumarin, hydrocoumarin, chroman-2-one, benzodihydropyrone, melilotin, melilotol, 2-chromanone, 1,2-benzodihydropyrone, melilotic lactone
PubChem CID 660
ChEBI CHEBI:16151
IUPAC Name 3,4-dihydrochromen-2-one
SMILES C1CC(=O)OC2=CC=CC=C21

Specifications

Melting Point 24°C to 25°C
Density 1.169
Boiling Point 272°C
Flash Point 130°C (266°F)
Refractive Index 1.556
Quantity 100 g
Beilstein 4584
Formula Weight 148.16
Percent Purity 99%
Chemical Name or Material Dihydrocoumarin
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 204-354-9
RTECSNumber MW5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only