missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dihydrocarvone, 98%, mixture of isomers

Catalog No. p-111209
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC203650250 25 g Glass bottle
AC203651000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AC203650250 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 203650250

CAS: 5524-05-0 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5524-05-0
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00001636
InChI Key AZOCECCLWFDTAP-RKDXNWHRSA-N
Synonym +-dihydrocarvone, d-dihydrocarvone, 1r,4r-dihydrocarvone, e-dihydrocarvone, unii-vta43h364z, 2r-trans-2-methyl-5-1-methylvinyl cyclohexan-1-one, 2r,5r-2-methyl-5-isopropenylcyclohexanone, 2r,5r-5-isopropenyl-2-methylcyclohexanone, 2r,5r-2-methyl-5-prop-1-en-2-yl cyclohexanone, cyclohexanone, 2-methyl-5-1-methylethenyl-, 2r,5r
PubChem CID 22227
ChEBI CHEBI:154
IUPAC Name (2R,5R)-2-methyl-5-prop-1-en-2-ylcyclohexan-1-one
SMILES CC1CCC(CC1=O)C(=C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.9255g/mL
Boiling Point 87.0°C to 88.0°C (6.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4700 to 1.4740
Quantity 25 g
Specific Gravity 0.9255
Specific Rotation Condition + 14.00 (25.00°C c=neat)
Specific Rotation + 14.00
Formula Weight 152.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (+)-Dihydrocarvone, 98%
Show More Show Less
EINECSNumber 226-872-4
TSCA TSCA

RUO – Research Use Only