missing translation for 'onlineSavingsMsg'
Learn More

Dihydrocapsiate, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013490
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013490 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013490 Supplier Merck Emd Millipore Supplier No. 0508825MG
Only null left
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 205687-03-2
Linear Formula C18H28O4
Synonym (4-Hydroxy-3-methoxyphenyl)methyl 8-methylnonanoate; 4-Hydroxy-3-methoxybenzyl 8-methylnonanoate; DCT; Vanillyl 8-methylnonanoate
Type Analytical Standard
Quantity 25 mg
Formula Weight 308.41
Percent Purity ≥95% (HPLC)
Grade Analytical Standard