missing translation for 'onlineSavingsMsg'
Learn More

Dihexylamine, 99+%

CAS: 143-16-8 | C12H27N | 185.35 g/mol

$104.65 - $104.65

Chemical Identifiers

CAS 143-16-8
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.35
MDL Number MFCD00009521
InChI Key PXSXRABJBXYMFT-UHFFFAOYSA-N
Synonym dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
PubChem CID 8920
IUPAC Name N-hexylhexan-1-amine
SMILES CCCCCCNCCCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC407591000
SDS View Documents
Thermo Scientific Chemicals
407591000
100 g Glass bottle
Each for $104.65
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

143-16-8
185.35
PXSXRABJBXYMFT-UHFFFAOYSA-N
8920
CCCCCCNCCCCCC
C12H27N
MFCD00009521
dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
N-hexylhexan-1-amine

Specifications

143-16-8
Colorless
236°C
Authentic
Glass bottle
1.4320 to 1.434
MFCD00009521
2927
0.8
Solubility in water: 0.3g/L(25°C)
CCCCCCNCCCCCC
185.35
185.35
Liquid
3°C
0.8000g/mL
104°C
99+%
C12H27N
(CH3(CH2)5)2NH
100 g
04,I,384
dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
PXSXRABJBXYMFT-UHFFFAOYSA-N
N-hexylhexan-1-amine
8920
99+%
Dihexylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 205-588-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only