missing translation for 'onlineSavingsMsg'
Learn More

Diglycolic Anhydride 98.0+%, TCI America™

Catalog No. D230825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D230825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D230825G Supplier TCI America Supplier No. D230825G

Chemical Identifiers

CAS 4480-83-5
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.072
MDL Number MFCD00006677
InChI Key PIYNUZCGMLCXKJ-UHFFFAOYSA-N
Synonym diglycolic anhydride, oxydiacetic anhydride, digylcolicanhydride, 4-maleimidobutyric acid hydrazide hydrochloride, r-1-methyl-2-pyrrolidineethanamine dihydrochloride, diglycolicanhydride, digylcolic anhydride, acetic acid, oxydi-, cyclic anhydride, p-dioxane-2,6-dione, pubchem24315
PubChem CID 78232
IUPAC Name 1,4-dioxane-2,6-dione
SMILES C1C(=O)OC(=O)CO1

Specifications

Melting Point 96°C
Color White
Boiling Point 130°C
Quantity 25 g
Formula Weight 116.07
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diglycolic Anhydride