missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Diglycolic acid

Catalog No. 1430731KG
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1430731KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1430731KG Supplier Sigma Aldrich Supplier No. 1430731KG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diglycolic acid

Specifications

Melting Point 140°C to 144°C (lit.)
CAS 110-99-6
Molecular Formula C4H6O5
Linear Formula O(CH2COOH)2
MDL Number MFCD00004309
Synonym 2,2 -Oxydiacetic acid
Molecular Weight (g/mol) 134.09
Percent Purity 98%
Quantity 1 kg