missing translation for 'onlineSavingsMsg'
Learn More

Diglycidyl ether, 95%, Thermo Scientific™

Chemical Identifiers

CAS 2238-07-5
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00080479
InChI Key GYZLOYUZLJXAJU-UHFFFAOYNA-N
IUPAC Name 2-{[(oxiran-2-yl)methoxy]methyl}oxirane
SMILES C(OCC1CO1)C1CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC383130010
SDS View Documents
Thermo Scientific Chemicals
383130010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC383130050
SDS View Documents
Thermo Scientific Chemicals
383130050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2238-07-5
130.14
GYZLOYUZLJXAJU-UHFFFAOYNA-N
C(OCC1CO1)C1CO1
C6H10O3
MFCD00080479
2-{[(oxiran-2-yl)methoxy]methyl}oxirane

Specifications

2238-07-5
260°C
Authentic
Glass bottle
1.4460 to 1.448
1g
GYZLOYUZLJXAJU-UHFFFAOYNA-N
2-{[(oxiran-2-yl)methoxy]methyl}oxirane
130.14
Diglycidyl ether
1.1200g/mL
64°C
94% min. (GC)
C6H10O3
MFCD00080479
1.12
C(OCC1CO1)C1CO1
130.14
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May form explosive peroxides.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

EINECSNumber : 218-802-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only