Perkin Elmer Canada DigiTUBEs 100 mL with RackLock, Includes Caps, Qty. 300
Catalog No. NC0806941
RackLock design allows for easy, one-handed closure of leak-free screw cap. All-in-one tubes eliminate the need for five separate vessels: Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RackLock design allows for easy, one-handed closure of leak-free screw cap. All-in-one tubes eliminate the need for five separate vessels: Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.