missing translation for 'onlineSavingsMsg'
Learn More

Digitonin, TCI America™

Catalog No. D0540100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0540100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0540100MG Supplier TCI America Supplier No. D0540100MG

Chemical Identifiers

CAS 11024-24-1
Molecular Formula C56H92O29
Molecular Weight (g/mol) 1229.32
MDL Number MFCD00077729
InChI Key UVYVLBIGDKGWPX-QYLWGQLPSA-N
Synonym digitonin
PubChem CID 102004607
IUPAC Name (2S,3R,4S,5S,6R)-2-{[(2S,3R,4S,5S,6R)-2-{[(2S,3R,4S,5R,6R)-2-{[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(1'R,2R,2'S,3'S,4'R,7'S,8'R,9'S,12'S,13'S,15'R,16'R,18'S)-5,7',9',13'-tetramethyl-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosane]-3',15'-dioloxy]oxan-3-yl]oxy}-5-hydroxy-6-(hydroxymethyl)-4-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}oxan-3-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C[C@H]1[C@H]2[C@@H](O[C@]11CCC(C)CO1)[C@@H](O)[C@H]1[C@@H]3CC[C@H]4C[C@@H](O[C@@H]5O[C@H](CO)[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O[C@@H]7OC[C@@H](O)[C@H](O)[C@H]7O)[C@H]6O[C@@H]6O[C@H](CO)[C@H](O)[C@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@H]6O)[C@H](O)[C@H]5O)[C@H](O)C[C@]4(C)[C@H]3CC[C@]21C

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 1229.32
Physical Form Crystalline Powder
Chemical Name or Material Digitonin
RTECSNumber IH2050050
TSCA Yes