Learn More
MilliporeSigma™ Digitonin Solution- high purity
Description
- Uses:
- Has been used to solubilize receptors and permeabilize membranes for sample preparation and analysis
- Removal of cholesterol and cellular membrane permeabilization
- Due to digitonin's mechanism of action, many other epigenetic applications utilize digitonin for nuclear extraction
Specifications
Specifications
| Content And Storage | 5 x 1 mL 2% Digitonin in DMSO. Store at -20°C |
For research purposes only