Learn More
Laxco™ Digital Upgrade Kit
GREENER_CHOICE
Catalog No. SLI3PDUKIT3
Change view
Click to view available options
For Use With (Equipment):
SLi3 PRO Series Microscopes
SLi6 PRO Series Microscopes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
SLI3PDUKIT3 SLi3 PRO Series Microscopes
SLI6PDUKIT2 SLi6 PRO Series Microscopes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SLI3PDUKIT3 Supplier Laxco™ Supplier No. SLI3PDUKIT3
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This Digital Upgrade Kit provides image capture capabilities to microscopes. Options for SLi3 PRO Series and SLi6 PRO Series Microscopes.
  • SeBaCamCool 1.7 C Cooled Color 1.7 Megapixel CMOS Camera
  • SeBalit Software-Fluorescent Image Capture Software
  • Mount adaptor included
  • Options available for use with SLi3 PRO Series Microscopes or SLi6 PRO Series Microscopes
Specifications
Includes Camera, Image Capture Software, Mount Adaptor
For Use With (Equipment) SLi3 PRO Series Microscopes
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Eyepiece WF 10X/22 mm, with rubber eyeguards
Lighting Type LED, Fluorescence
Head Style Trinocular
Camera Type CMOS
Display Type Computer
Product Type Educational Microscope
Resolution 1.7 MP