Learn More
Mopec™ Digital Tissue Flotation Bath Lid
Catalog No. 22445367
Change view
Click to view available options
Type:
Flotation Bath Lid
Catalog No. Type
22445367 Flotation Bath Lid
1 options
Catalog No. 22445367 Supplier Mopec™ Supplier No. BK150L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Digital Tissue Flotation Bath features a high-contrast background that allows users to better view tissue sections while also detecting foreign particles floating on the surface of the water.
  • Lid for the Mopec™ BK150 Digital Tissue Flotation Bath
Specifications
For Use With (Equipment) Mopec Digital Tissue Flotation Bath
Temperature Range 30°C to 70°C
Type Flotation Bath Lid
Electrical Requirements None