Learn More
Fisherbrand™ Digital Thermometers with Stainless-Steel Probe on Cable
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas
 View more versions of this product
Catalog No. 150779D
Only null left
Description
Description
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas. Fisher Scientific™ Digital thermometer with stainless-steel probe is chemically resistant to most acids, bases and solvents and may be used to measure temperature of gases, liquids, food, and other semisolids.
  • Models 15-077-16, -15, -21, -8B, -29 and 14-648-46 have alarm that can be set to alert user to temperatures that exceed setpoints
  • Some models (such as 15-077-16 and -15) have special features that print temperature, high/low memory and time of day; 15-077-29 thermometer has additional timer alarms
  • For a miniature-size thermometer, see model 15-077-32
For use in cuvets, soil, incubators, petroleum products and piercing food samples. Waterproof thermometers (models 15-077-9E, -9D and 14-648-46) are ideal for wet labs, wash-down areas and outdoor applications in the agricultural, petrochemical, food, and pharmaceutical industries.
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Chemical Identifiers
ISO 17025, A2LA, NIST
Traceable Digital Thermometer w/Probe
Yes
Digital LCD
Specifications
ISO 17025, A2LA, NIST
Wet labs, wash down areas and outdoor applications in the agricultural, petrochemical, food and pharmaceutical industries
1.3 cm
120 in.
127 mm
12.7 cm
0.1°C
56.6 g
5 in.
300°C
-50°C
N/A
±1°C
9 cm
0.12 in.
0.5 in.
Suction cup, magnets, Velcro™, traceable certificate and 1.5V Silver Oxide battery
5 in.
304.8 cm
Plastic
2 oz.
Traceable Digital Thermometer w/Probe
572°F
-58°F
-58°F to +572°F
Yes
Digital LCD
Product Suggestions
Product Suggestions