missing translation for 'onlineSavingsMsg'
Learn More

OHAUS Digital MicroPlate Genie Pulse

Catalog No. 01804336
Change view
Click to view available options
Product Type:
Microplate Mixer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
01804336 Microplate Mixer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01804336 Supplier OHAUS Supplier No. 83041396

Precise speed control or pulse vortexing with 1.0 mm diameter orbit.

  • A true pulsing action for a more random motion and thorough mixing.
  • Fully programmable time, and Run/Stop modes.

Specifications

Orbit 1 mm
Speed Range 500 to 3000 rpm
Voltage 120 V
Product Type Microplate Mixer
Model Digital MicroPlate Genie Pulse