LABORATORY INCUBATOR, LARGE CAPACITY, 38.6 CU FT, SOLID DOOR w/ VIEW, ROLL-IN FLOOR, OUTLET, ACCESS PORT, 115V This incubator provides extra large capacities while minimizing the amount of floor space needed. The temperature in this large space is kept uniform by integrating a highly responsive microprocessor with an appropriately sized heating element. An independent secondary temperature controller offers the added security of over temperature protection. Our unique forced air circulation system creates a one-pass circulation pattern that provides both exceptional temperature uniformity and rapid heat recovery. The chamber floors are ruggedly reinforced to support roller apparatus or shakers. The SMI39 is supplied with six sturdy shelves that will not sag or bend under heavy loads. More information available at www.shellab.com