Learn More
Research Products Corporation Traceable Digital Countdown Timer
Catalog No. 501367786
Encompass
Large 1'' display offers easy viewing from across any laboratory. Features reliable 3 key operation with countdown setting in 1 second increments. Counts down from 99 minutes, 59 seconds, alarms and counts up during alarming to indicate elapsed time. Automatic Bounceback Memory allows for repetitive timing.
Features 4 way holder: hanger, clip-on, easel stand and magnetic back. Supplied with certificate of calibration and battery.
Catalog No. 501367786 Supplier Research Products Corporation Supplier No. 805135
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Large 1'' display offers easy viewing from across any laboratory. Features reliable 3 key operation with countdown setting in 1 second increments. Counts down from 99 minutes, 59 seconds, alarms and counts up during alarming to indicate elapsed time. Automatic Bounceback Memory allows for repetitive timing.
Features 4 way holder: hanger, clip-on, easel stand and magnetic back. Supplied with certificate of calibration and battery.