missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotators

Catalog No. 88882015
Change view
Click to view available options
Plug Type:
US Plug
Certifications/Compliance:
cCSAus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Certifications/Compliance
88882015 US Plug cCSAus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88882015 Supplier Thermo Scientific™ Supplier No. 88882015

missing translation for 'validMainframeMsg'

Requires

Minimum 1 drum order together with Rotator

The Digital Cel-Gro Tissue Culture Rotator has a brushless no-maintenance motor that provides a gentle rotating motion for optimum results.

The Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator precisely mixes test tube contents. Achieve optimum culture results for tissue and virus growth, hormone production, and sample testing of viruses and cytotoxicity assays.

  • Digital display of Speed and Time
  • Speed range: 5 – 70 rpm
  • Speed Accuracy: ±1 rpm
  • Adjustable rocking Angle: 95°- 180°
  • Loading Capacity: 5 kg
  • Timer Range: 0 - 9999mins
  • Optional multiple size drums offering different capacity of tube rotation

Order Info

Please order minimum 1 drum together with the rotator

Warranty and Services

2 years

Specifications

Rotating Angle 90°
Load Bearing Capacity (Metric) 5 kg
Speed Control Accuracy ±1 rpm
Tilt Angle 95 to 180°
Speed Range 5 to 70 rpm
Electrical Requirements 100/240 V
Voltage 100/240 V
Temperature Range (Metric) 5°C to 40°C
Amperage 0.2 A
Product Type Tissue Culture Rotator
Certifications/Compliance cCSAus
Plug Type US Plug
Frequency 50/60 Hz
Material Stainless Steel Platform
Dimensions (L x W x H) 383 x 260 x 522 mm (15.1 x 10.2 x 20.6 in.)
Shipping Weight (English) 26 lb.
Shipping Weight (Metric) 11.8 kg
Alarms Audible alarm
Controller Type Digital control
Model Digital Cel-Gro Tissue Culture Rotator
Display Digital
For Use With (Application) Cell Culture
For Use With (Equipment) Accessories: 88882149, 88882150,88882151, 88882152, 88882153, 88882154, 88882155, 88882156, 88882157
Humidity Range 20 to 85%
Includes Equipment only
Timer Range 0 to 9999 min.
Warranty and Service Offering 2 Years
Show More Show Less