missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digestion Reagent, with Mercury Catalyst, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Digestion Reagent |
| Quantity | 1 L |
| Color | Colorless |
| Name Note | With Mercury Catalyst |
| CAS Min % | 65.0, 24.42, 8.45, 0.13 |
| Packaging | Natural Poly Bottle |
| CAS Max % | 68.0, 25.42, 8.8, 0.14 |
| CAS | 7664-93-9, 7778-80-5, 21908-53-2 |