Rice Lake Weighing System DIG BABY SCALE 99 X .1LB
Catalog No. 50597138
Medical Scale, Rice Lake Weighing Systems, Hanging OS-45 45kg x 0.05kg/99lb x 0.1lb/99lb x 1oz Includes carrying case, (2) AA Batteries, bottom hook fully attached, top hook removable, adjustable auto shutoff, units kg/lb/lb-oz
Catalog No.
50597138
Supplier
Rice Lake Weighing SystemSupplier No.
130294
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Medical Scale, Rice Lake Weighing Systems, Hanging OS-45 45kg x 0.05kg/99lb x 0.1lb/99lb x 1oz Includes carrying case, (2) AA Batteries, bottom hook fully attached, top hook removable, adjustable auto shutoff, units kg/lb/lb-oz