missing translation for 'onlineSavingsMsg'
Learn More

Difurfurylsulfide, 96%, Thermo Scientific™

Chemical Identifiers

CAS 13678-67-6
Molecular Formula C10H10SO2
Molecular Weight (g/mol) 194.25
InChI Key UYLKDZXJEKFFHJ-UHFFFAOYSA-N
Synonym difurfuryl sulfide, furfuryl sulfide, difurfurylsulfide, 2-furfuryl monosulfide, furan, 2,2'-thiobis methylene bis, bis 2-furfuryl sulfide, unii-x6o686cpco, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, 2-difurfuryl sulfide
PubChem CID 61659
IUPAC Name 2-(furan-2-ylmethylsulfanylmethyl)furan
SMILES C1=COC(=C1)CSCC2=CC=CO2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC265080050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13678-67-6
194.25
difurfuryl sulfide, furfuryl sulfide, difurfurylsulfide, 2-furfuryl monosulfide, furan, 2,2'-thiobis methylene bis, bis 2-furfuryl sulfide, unii-x6o686cpco, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, 2-difurfuryl sulfide
2-(furan-2-ylmethylsulfanylmethyl)furan
C10H10SO2
UYLKDZXJEKFFHJ-UHFFFAOYSA-N
61659
C1=COC(=C1)CSCC2=CC=CO2

Specifications

13678-67-6
>110°C
95% min. (GC)
1.5540 to 1.5560
difurfuryl sulfide, furfuryl sulfide, difurfurylsulfide, 2-furfuryl monosulfide, furan, 2,2'-thiobis methylene bis, bis 2-furfuryl sulfide, unii-x6o686cpco, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, 2-difurfuryl sulfide
UYLKDZXJEKFFHJ-UHFFFAOYSA-N
2-(furan-2-ylmethylsulfanylmethyl)furan
61659
96%
135.0°C to 143.0°C (14.0mmHg)
Authentic
C10H10SO2
17,III,355
Solubility in water: insoluble
C1=COC(=C1)CSCC2=CC=CO2
194.25
194.25
Difurfurylsulfide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 237-172-3

SDS
Documents

Documents

RUO – Research Use Only