missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Difurfuryl Sulfide 98.0+%, TCI America™
$68.97 - $206.90
Chemical Identifiers
| CAS | 13678-67-6 |
|---|---|
| Molecular Formula | C10H10O2S |
| Molecular Weight (g/mol) | 194.248 |
| MDL Number | MFCD00010081 |
| InChI Key | UYLKDZXJEKFFHJ-UHFFFAOYSA-N |
| Synonym | difurfuryl sulfide, furfuryl sulfide, difurfurylsulfide, 2-furfuryl monosulfide, furan, 2,2'-thiobis methylene bis, bis 2-furfuryl sulfide, unii-x6o686cpco, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, 2-difurfuryl sulfide |
| PubChem CID | 61659 |
| IUPAC Name | 2-(furan-2-ylmethylsulfanylmethyl)furan |
| SMILES | C1=COC(=C1)CSCC2=CC=CO2 |
Chemical Identifiers
| 13678-67-6 | |
| 194.248 | |
| UYLKDZXJEKFFHJ-UHFFFAOYSA-N | |
| 61659 | |
| C1=COC(=C1)CSCC2=CC=CO2 |
| C10H10O2S | |
| MFCD00010081 | |
| difurfuryl sulfide, furfuryl sulfide, difurfurylsulfide, 2-furfuryl monosulfide, furan, 2,2'-thiobis methylene bis, bis 2-furfuryl sulfide, unii-x6o686cpco, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, 2-difurfuryl sulfide | |
| 2-(furan-2-ylmethylsulfanylmethyl)furan |
Specifications
| 13678-67-6 | |
| 143°C | |
| MFCD00010081 | |
| difurfuryl sulfide, furfuryl sulfide, difurfurylsulfide, 2-furfuryl monosulfide, furan, 2,2'-thiobis methylene bis, bis 2-furfuryl sulfide, unii-x6o686cpco, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, 2-difurfuryl sulfide | |
| C1=COC(=C1)CSCC2=CC=CO2 | |
| 194.248 | |
| 194.25 | |
| Crystalline Lumps |
| White-Yellow | |
| C10H10O2S | |
| 5 g | |
| UYLKDZXJEKFFHJ-UHFFFAOYSA-N | |
| 2-(furan-2-ylmethylsulfanylmethyl)furan | |
| 61659 | |
| ≥98.0% (GC) | |
| Difurfuryl Sulfide |
Safety and Handling
TSCA : Yes