missing translation for 'onlineSavingsMsg'
Learn More

(Difluoromethyl)trimethylsilane 97.0+%, TCI America™

Catalog No. p-7128243
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43641ML 1 mL
D43645ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43641ML Supplier TCI America Supplier No. D43641ML
Only null left

Chemical Identifiers

CAS 65864-64-4
Molecular Formula C4H10F2Si
Molecular Weight (g/mol) 124.206
MDL Number MFCD19381658
InChI Key OOKFLLNDYNWCHK-UHFFFAOYSA-N
PubChem CID 11170968
IUPAC Name difluoromethyl(trimethyl)silane
SMILES C[Si](C)(C)C(F)F

Specifications

Color Yellow
Boiling Point 52°C
Quantity 1 mL
UN Number 1993
Formula Weight 124.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (Difluoromethyl)trimethylsilane
TSCA No
Recommended Storage Refrigerator