missing translation for 'onlineSavingsMsg'
Learn More

(Difluoromethyl)benzene 98.0+%, TCI America™

Catalog No. p-7128546
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49151G 1 g
D49155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49151G Supplier TCI America Supplier No. D49151G
Only null left

Chemical Identifiers

CAS 455-31-2
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.12
MDL Number MFCD03788488
InChI Key JDZLOJYSBBLXQD-UHFFFAOYSA-N
Synonym alpha,alpha-Difluorotoluene
PubChem CID 640452
IUPAC Name (difluoromethyl)benzene
SMILES FC(F)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 135°C
Quantity 1 g
UN Number 2920
Formula Weight 128.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Difluoromethyl)benzene