missing translation for 'onlineSavingsMsg'
Learn More

Difluoromethyl 2-Pyridyl Sulfone 98.0+%, TCI America™

Catalog No. D53901G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D53901G Supplier TCI America Supplier No. D53901G
Only null left

Chemical Identifiers

CAS 1219454-89-3
Molecular Formula C6H5F2NO2S
Molecular Weight (g/mol) 193.17
MDL Number MFCD17010193
InChI Key YRQNSTAWTLXCEZ-UHFFFAOYSA-N
Synonym 2-(Difluoromethylsulfonyl)pyridine, 2-PySO2CF2H
PubChem CID 46188280
IUPAC Name 2-difluoromethanesulfonylpyridine
SMILES FC(F)S(=O)(=O)C1=CC=CC=N1

Specifications

Melting Point 49°C
Color White
Quantity 1 g
Formula Weight 193.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Difluoromethyl 2-Pyridyl Sulfone