missing translation for 'onlineSavingsMsg'
Learn More

Difluoroacetic Acid 97.0+%, TCI America™

Catalog No. p-7126723
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14235G 5 g
D142325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D14235G Supplier TCI America Supplier No. D14235G
Only null left

Chemical Identifiers

CAS 381-73-7
Molecular Formula C2H2F2O2
Molecular Weight (g/mol) 96.033
MDL Number MFCD00004220
InChI Key PBWZKZYHONABLN-UHFFFAOYSA-N
Synonym difluoroacetic acid, acetic acid, difluoro, difluoressigsaeure, 1,1-difluoroacetic acid, unii-zqk1c95k3n, zqk1c95k3n, acetic acid, 2,2-difluoro, difluoroaceticacid, difluoracetic acid, difluoro-acetic acid
PubChem CID 9788
ChEBI CHEBI:23716
IUPAC Name 2,2-difluoroacetic acid
SMILES C(C(=O)O)(F)F

Specifications

Melting Point 0°C
Boiling Point 135°C
Quantity 5 g
UN Number 3265
Formula Weight 96.03
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Difluoroacetic Acid
RTECSNumber AG9900000
TSCA Yes