missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Difluoroacetaldehyde Ethyl Hemiacetal 90.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 126.10 |
| Percent Purity | ≥90.0% (NMR) |
| Physical Form | Liquid |
| Chemical Name or Material | Difluoroacetaldehyde Ethyl Hemiacetal |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |