missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Trypticase™ Soy Agar

Catalog No. B11043
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11043 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11043 Supplier BD Supplier No. 211043
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use with or without blood for the cultivation and isolation of a variety of organisms

Specifications

Color Light beige
Form Solid
For Use With (Application) Used with or without Blood for the cultivation and isolation of a variety of organism
Quantity 500 g