missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Sabouraud Dextrose Broth

Catalog No. DF0382179
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0382179 500 g Bottle
DF0382071 2 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0382179 Supplier BD Supplier No. 238230
Edge

Used in qualitative procedures for culturing yeasts, molds and aciduric microorganisms. BD™ Difco™ Sabouraud Dextrose Broth meets United States Pharmacopeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP) performance specifications.

Sabouraud Dextrose Broth is used for culturing yeasts and molds in cosmetics. The USP recommends the use of Sabouraud Dextrose Broth when isolating Candida albicans from nonsterile pharmaceutical products.

  • Sabouraud dextrose media are peptone media supplemented with dextrose to support the growth of fungi
  • Peptones are sources of nitrogenous growth factors
  • The carbohydrate provides an energy source for the growth of microorganisms
  • pH (3.0% solution, 25°C): 5.6 ± 0.2
  • Displays cultural response to: Aspergillus brasiliensis (niger) 16404, Candida albicans 10231, Lactobacillus casei 9595, Saccharomyces cerevisiae 9763, Candida albicans 10231

Specifications

Certifications/Compliance BAM, EP, JP, USP
Color Light beige
Content And Storage 2 to 25°C
Form Dehydrated Powder
Media Type Sabouraud Dextrose Broth
Product Type Microbiology Media
Formulation Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 5g/L, Dextrose 20g/L
For Use With (Application) Cultivation of Yeasts, molds and aciduric microorganisms
Packaging Type Bottle
Quantity 500 g