missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Sabouraud Dextrose Agar

Catalog No. DF0109171
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0109171 500 g Bottle
DF0109082 10 kg Pail
DF0109073 2 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. DF0109171 Supplier BD Supplier No. 210950
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rendered more selective for fungi by the addition of antimicrobics. BD™ Difco™ Sabouraud Dextrose Agar is a general-purpose medium used in the cultivation of yeasts, molds and aciduric microorganisms. It is also recommended for the testing of cosmetics and food.

Difco™ Sabouraud Dextrose Agar is a dehydrated culture media used in qualitative procedures for cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes.

  • The addition of antimicrobics is a modification designed to increase bacterial inhibition.
  • The low pH of approximately 5.6 is favorable for the growth of fungi and slightly inhibitory to contaminating bacteria in clinical specimens.
  • General Chapters 61 and 62 of the USP describe test methods for using Sabouraud Dextrose Agar when performing the microbial enumeration tests and tests for isolating Candida albicans from nonsterile pharmaceutical products.

Specifications

Certifications/Compliance BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Color Beige
Content And Storage 2 to 25°C
Form Dehydrated
Media Type Sabouraud Dextrose Agar
Product Type Agar
Formulation Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 5g/L, Dextrose 40g/L, Agar 15g/L
For Use With (Application) Cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes
Packaging Type Bottle
Quantity 500 g