missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Difco™ PPLO Broth
Used for isolating and cultivating Mycoplasma, when supplemented with nutritive enrichments.
Supplier: Gibco™ 255410
Description
A complete media used in the cultivation of microorganisms from the class Mollicutes. Used in animal health applications for the propagation of Mycoplasma species such as Mycoplasma mycoides, Mycoplasma gallisepticum, and others for vaccine production.Specifications
| Storage conditions: 2°C to 25°C. Â Store in cool, dry conditions in well-sealed containers. | |
| Isolating and cultivating Mycoplasma | |
| 10 kg |
| Dehydrated culture media | |
| Difco™ | |
| Powder |